Learn More
Medchemexpress LLC N-biotinyl-1,6-hexanediamine | 65953-56-2 | 99.6% | 342.5 | C16H30N4O2S | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW343597250MG
Encompass_Preferred
N-biotinyl-1,6-hexanediamine is a biotinylated reagent used for labeling, detection, and affinity-capture workflows in biochemical research. The molecule features a biotin moiety linked by a six-carbon spacer to a terminal primary amine, enabling versatile conjugation to biomolecules and surface chemistries. Offered in multiple package sizes and high purity suitable for laboratory use.
  • Provides a biotin handle for conjugation and detection.
  • Six-carbon spacer to reduce steric hindrance in conjugates.
  • Terminal primary amine enables common coupling chemistries.
  • High purity suitable for analytical and preparative experiments.
  • Available in multiple package sizes for flexibility in use.
Catalog No. 50-002-82034