Learn More
Medchemexpress LLC N-biotinyl-6-aminohexanoic acid | 72040-64-3 | MFCD00144853 | 99.8% | 357.47 g/mol | C16H27N3O4S | 250 MG
SDP
Supplier:  Medchemexpress LLC HY101218250MG
Encompass_Preferred
N-biotinyl-6-aminohexanoic acid is a biotin-conjugated aminohexanoic acid reagent used to introduce a biotin tag onto proteins, peptides, and other biomolecules for affinity capture, detection, or immobilization in research applications. It is supplied as a white to off-white solid with high purity and is suitable for analytical and preparative laboratory workflows.
  • Provides a biotin handle for affinity capture and detection.
  • High purity (~99.8%) suitable for research use.
  • White to off-white solid, easy to weigh and handle.
  • Compatible with streptavidin and avidin affinity systems.
  • Stable at room temperature for shelf storage; extended stability in solvent when kept at low temperatures.
Catalog No. 50-002-12933