Learn More
Medchemexpress LLC N-Boc-4-piperidinemethanol | 123855-51-6 | MFCD02094488 | 99.9% | 215.29 | 1 ML

Supplier: Medchemexpress LLC HY2162910MM/1ML
N-Boc-4-piperidinemethanol, also known as tert-butyl 4-(hydroxymethyl)piperidine-1-carboxylate, is a chemical compound with the molecular formula C11H21NO3 and a molecular weight of 215.29. It is commonly used in laboratory settings and has a high purity of 99.91%.
- White to yellow solid appearance.
- Molecular formula C11H21NO3.
- Molecular weight of 215.29.
- Purity (GC) of 99.91%.
- Consistent with structure by 1H NMR Spectrum.
- May cause skin irritation.
- May cause serious eye irritation.
- May cause respiratory irritation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.