Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Boc-5-bromoindole | 182344-70-3 | MFCD02090067 | 100.0% | 296.16 g/mol | C13H14BrNO2 | 25 G
SDP

Supplier:  Medchemexpress LLC 25GHYW021597

Encompass_Preferred

N-Boc-5-bromoindole is a Boc-protected indole derivative used as a synthetic intermediate for the preparation of di-Boc-protected 5-aminoindole via Buchwald-Hartwig amination followed by regioselective bromination. It is supplied as a high-purity solid for medicinal chemistry and organic synthesis research.

  • High reported purity (99.98%).
  • Molecular weight 296.16 g/mol.
  • Chemical formula C13H14BrNO2.
  • Suitable as a building block for cross-coupling and amination reactions.
  • Available in multiple pack sizes including 25 g.

Catalog No. 50-003-03677


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.