Learn More
Medchemexpress LLC N-Boc-5-bromoindole | 182344-70-3 | MFCD02090067 | 100.0% | 296.16 g/mol | C13H14BrNO2 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0215975G
Encompass_Preferred
N-Boc-5-bromoindole is a Boc-protected, brominated indole used as a synthetic intermediate in organic chemistry. It is commonly employed to prepare di-Boc-protected 5-aminoindole derivatives via Buchwald-Hartwig amination of the indole N-position with tert-butyl carbamate, followed by regioselective bromination and downstream functionalization.
  • Boc-protected indole suitable for N-functionalization.
  • Enables Buchwald-Hartwig amination workflows.
  • Compatible with palladium-catalyzed cross-coupling reactions.
  • High purity for reliable synthetic outcomes.
  • Available in small pack sizes for research-scale synthesis.
  • Defined molecular formula and molar mass for accurate calculations.
Catalog No. 50-003-01344