Learn More
Medchemexpress LLC N-Boc-D-proline | 37784-17-1 | ≥98.0% | 215.25 | 100 G

Supplier: Medchemexpress LLC HY40115100G
Boc-D-proline is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
- Purity: 98.0%
- For research use only
- Appearance: Solid
- Color: White to off-white
- Shipping: Room temperature in continental US; may vary elsewhere
- Storage: Powder: -20°C for 3 years; 4°C for 2 years; in solvent: -80°C for 6 months; -20°C for 1 month
- Soluble in DMSO (200 mg/mL)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.