Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Boc-diethanolamine | 103898-11-9 | MFCD01862953 | 100.0% | 205.25 g/mol | C9H19NO4 | 50 G
SDP

Supplier:  Medchemexpress LLC HYW04407850G

Encompass_Preferred

N-Boc-diethanolamine is a Boc-protected diethanolamine reagent used as a cleavable linker in the synthesis of proteolysis targeting chimeras (PROTACs) and antibody-drug conjugates (ADCs). It is supplied with analytical and safety documentation for research use.

  • Boc-protected diethanolamine suitable for linker chemistry.
  • High purity with analytical data available.
  • Molecular formula C9H19NO4, molecular weight 205.25 g/mol.
  • Available in multiple pack sizes, including a 50 g laboratory pack.
  • Downloadable data sheet, SDS, COA, H NMR, and GC files provided.
  • Intended for research use only; not for human consumption.

Catalog No. 50-002-03196


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.