Learn More
Medchemexpress LLC N-BOC-DL-serine methyl ester | 69942-12-7 | 219.23 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW005308500G
Encompass_Preferred
N-BOC-DL-serine methyl ester is a serine derivative. It is utilized in various synthetic processes, including the synthesis of α,β-dehydro-α-amino acids, as well as anti-cancer agents such as quinazolinone derivatives that inhibit PI3K activity. It also plays a role in the synthesis of tricyclic pyrolopyranopyridines, which are known to inhibit protein kinase activity.
  • Used for synthesis of α,β-dehydro-α-amino acid
  • Used for synthesis of anti-cancer agents (quinazolinone derivatives inhibiting PI3K activity)
  • Used for synthesis of tricyclic pyrolopyranopyridines (inhibiting protein kinase activity)
  • Store in pure form at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-45991