Learn More
Medchemexpress LLC N-Boc-dolaproine | 120205-50-7 | 100.0% | 287.36 | 1 G
SDP
Supplier:  Medchemexpress LLC HY330461G
Encompass_Preferred
N-Boc-dolaproine ((2R,3R)-BOC-dolaproine) is an amino acid residue of the pentapeptide Dolastatin 10. Dolastatin 10 inhibits tubulin polymerization and mitosis and possesses anticancer activity. This product is a key intermediate utilized in the synthesis of Dolastatin 10.
  • Amino acid residue of Dolastatin 10
  • Inhibits tubulin polymerization and mitosis
  • Possesses anticancer activity
  • Key intermediate for synthesis of Dolastatin 10
  • Colorless to light yellow oil
  • Ships at room temperature in continental US
  • Store at 4°C under nitrogen
Catalog No. 50-004-03542