Learn More
Medchemexpress LLC N-Boc-dolaproine | 120205-50-7 | 99.96% | 287.36 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY33046100MG
Encompass_Preferred
N-Boc-dolaproine is an amino acid residue of the pentapeptide Dolastatin 10. Dolastatin 10 inhibits tubulin polymerization and mitosis and possesses anticancer activity. N-Boc-dolaproine is a key intermediate used in the synthesis of Dolastatin 10.
  • Appearance: Oil
  • Color: Colorless to light yellow
Catalog No. 50-004-04724