Learn More
Medchemexpress LLC N-Boc-dolaproine-amide-Me-Phe | 3026596-72-2 | 98.46% | C23H36N2O4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY4905310MM/1ML
Encompass_Preferred
N-Boc-dolaproine-amide-Me-Phe is an amino acid residue derived from Dolastatin 10, a pentapeptide. Dolastatin 10 is recognized for its ability to inhibit tubulin polymerization and mitosis, and it demonstrates anticancer activity.
  • Amino acid residue of Dolastatin 10
  • Inhibits tubulin polymerization
  • Inhibits mitosis
  • Exhibits anticancer activity
  • For research use only
Catalog No. 50-003-58125