Learn More
Medchemexpress LLC N-Boc-L-Prolinal | 69610-41-9 | 98.8% | C10H17NO3 | 10 G
SDP
Supplier:  Medchemexpress LLC HY7892710G
Encompass_Preferred
N-Boc-L-Prolinal is a proline derivative featuring a Boc protecting group. This compound serves as a valuable building block for synthesizing biologically active peptide mimetics.
  • Proline derivative featuring a Boc protecting group.
  • Serves as a valuable building block for synthesizing biologically active peptide mimetics.
  • Utilized in the synthesis of Dolastatin 10 analogs, which exhibit anti-colon cancer activity.
Catalog No. 50-004-32073