Learn More
Medchemexpress LLC N-Boc-MeVal | 1268729-59-4 | 245.32 | 1 G

Supplier: Medchemexpress LLC 1GHY41062
N-Boc-MeVal is an ADC linker with a BOC protecting group. This product is suitable for applications targeting Antibody-drug Conjugates.
- Targeted for ADC linker applications
- BOC protecting group included
- Store at 4°C, sealed, away from moisture and light
- For solutions, store at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.