Learn More
Medchemexpress LLC N-Boc-N-bisPEG2-OH 100mg | 275385-03-0 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY117079100MG
Encompass_Preferred
N-Boc-N-bis(PEG2-OH) is a Boc-protected, PEG-based bifunctional linker bearing two PEG2 (diethylene glycol) hydroxyl termini. It is used in the synthesis of PROTACs and as a cleavable linker for antibody-drug conjugates (ADCs), and is supplied for research applications as a colorless to light-yellow liquid.
  • Boc-protected nitrogen enables orthogonal deprotection and downstream coupling reactions.
  • Two PEG2 hydroxyl termini increase solubility and provide flexible spacer length.
  • Applicable to PROTAC assembly and cleavable ADC linker chemistry.
  • Molecular weight 293.36; chemical formula C13H27NO6; CAS 275385-03-0.
  • Available in small research quantities, e.g., 100 MG, for laboratory synthesis workflows.
Catalog No. 50-002-29238