Learn More
Medchemexpress LLC N-Boc-N-bisPEG2-OH 1g | 275385-03-0 | 293.36 g·mol⁻1 | C13H27NO6 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1170791G
Encompass_Preferred
N-Boc-N-bis(PEG2-OH) is a Boc-protected bis(PEG2) diol used as a linker building block in PROTAC and antibody-drug conjugate synthesis. It is a small, polar bifunctional molecule (molecular weight 293.36 g·mol⁻1) supplied as a solid and accompanied by a data sheet and safety data sheet for handling and storage.
  • Boc-protected bis-PEG2 diol linker suitable for PROTAC and ADC chemistry.
  • Small, polar building block with molecular weight 293.36 g·mol⁻1.
  • Available as a solid in 1 g pack size.
  • Provided with a data sheet and safety data sheet for handling and storage.
  • Two terminal hydroxy groups for chemoselective conjugation.
Catalog No. 50-003-08342