Learn More
Medchemexpress LLC N-Boc-N-bisPEG2-OH 250mg | 275385-03-0 | 250 MG
SDP
Supplier:  Medchemexpress LLC HY117079250MG
Encompass_Preferred
N-Boc-N-bis(PEG2-OH) is a PEG/alkyl/ether-based bifunctional linker used to assemble ADCs and PROTACs. Supplied as a 250 mg vial, it contains an N-Boc-protected nitrogen and two PEG2-derived hydroxyl termini that enable conjugation and subsequent functionalization in medicinal chemistry workflows.
  • PEG-based bifunctional linker for conjugation chemistry.
  • N-Boc-protected amine for orthogonal deprotection strategies.
  • Two terminal hydroxyl groups for further functionalization.
  • Molecular formula C13H27NO6; molecular weight 293.36 g·mol⁻1.
  • Designed for linker assembly in ADC and PROTAC synthesis.
Catalog No. 50-002-29241