Learn More
Medchemexpress LLC N-Boc-O-tosyl hydroxylamine | 105838-14-0 | 287.33 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00043810G
Encompass_Preferred
N-Boc-O-tosyl hydroxylamine is a safe and efficient nitrogen source for the N-amination of aryl and alkyl amines. It facilitates the electrophilic N-amination of these amines, leading to the generation of β-Boc-protected aryl hydrazines and alkyl hydrazines.
  • Serves as a safe and efficient nitrogen source
  • Facilitates electrophilic N-amination of aryl amines
  • Facilitates electrophilic N-amination of alkyl amines
  • Leads to the generation of β-Boc-protected aryl hydrazines
  • Leads to the generation of β-Boc-protected alkyl hydrazines
  • Intended for research use only
Catalog No. 50-004-46621