Learn More
Medchemexpress LLC N-Boc-PEG1-bromide | 164332-88-1 | 97.9% | 268.15 g/mol | C9H18BrNO3 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY130503500MG
Encompass_Preferred
N-Boc-PEG1-bromide is a Boc-protected polyethylene glycol (PEG1) reagent bearing a terminal bromide, used as a short PEG spacer and alkylating reagent in the synthesis of proteolysis-targeting chimeras (PROTACs) and antibody-drug conjugates (ADCs) for research applications. Typical purity is 97.87% and molecular weight is 268.15 g/mol (CAS 164332-88-1).
  • Boc-protected polyethylene glycol reagent with a terminal bromide for alkylation.
  • Short PEG spacer (PEG1) to increase solubility and provide spacing.
  • Suitable for ADC and PROTAC linker synthesis.
  • Typical purity around 97.9% for research use.
  • Available as a 500 mg solid package for laboratory synthesis.
Catalog No. 50-002-03237