Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Boc-PEG10-alcohol | 2238844-74-9 | 557.67 | C25H51NO12 | 25 MG
SDP

Supplier:  Medchemexpress LLC HYW19085725MG

Encompass_Preferred

N-Boc-PEG10-alcohol is a polyethylene glycol (PEG)-based linker used in the synthesis of proteolysis-targeting chimeras (PROTACs). It combines a Boc-protected amino group with a terminal alcohol and a PEG10 spacer to provide flexibility and solubility when conjugating ligands for target proteins and E3 ubiquitin ligases. Intended for research use and bioconjugation applications.

  • Provides a flexible PEG10 spacer for linker design.
  • Contains a Boc-protected amino group and a terminal alcohol for orthogonal conjugation.
  • Molecular weight 557.67 and formula C25H51NO12.
  • Supplied in small research pack sizes suitable for synthesis (for example, 25 mg).
  • Intended for research use only; not for therapeutic use.

Catalog No. 50-002-83074


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.