Learn More
Medchemexpress LLC N-Boc-PEG10-alcohol | 2238844-74-9 | ≥98.0% | 557.67 g·mol⁻1 | C25H51NO12 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW19085710MG
Encompass_Preferred
N-Boc-PEG10-alcohol is a Boc-protected polyethylene glycol linker composed of ten ethylene glycol units terminating in a primary alcohol. It is intended for use as a flexible, hydrophilic spacer in PROTACs, ADCs, and other bioconjugation applications where controlled spacing and orthogonal protection are required. Molecular formula C25H51NO12; molecular weight 557.67 g·mol⁻1.
  • Boc-protected amine allows orthogonal deprotection and further functionalization.
  • Ten-unit PEG spacer increases solubility and provides flexible linker length.
  • Terminal hydroxyl enables diverse conjugation chemistries.
  • Suitable for PROTAC and ADC linker synthesis and other bioconjugations.
  • Supplied in small, research-scale quantities for synthetic workflows.
Catalog No. 50-002-83188