Learn More
Medchemexpress LLC N-Boc-PEG10-alcohol | 2238844-74-9 | 98.0% | 557.67 g/mol | C25H51NO12 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW190857100MG
Encompass_Preferred
N-Boc-PEG10-alcohol is a polyethylene glycol-based PROTAC linker used to connect target-binding ligands and E3 ligase ligands in Proteolysis Targeting Chimera (PROTAC) design. It provides a flexible, hydrophilic spacer to optimize inter-ligand distance and solubility during PROTAC assembly. Molecular weight 557.67 g/mol; CAS 2238844-74-9.
  • Provides a flexible, hydrophilic spacer for PROTAC linkers.
  • Enables linker length optimization to tune degradation potency.
  • Compatible with common conjugation chemistries.
  • Available in small-scale research quantities for synthesis.
  • Supplied as a solid suitable for storage and handling.
Catalog No. 50-002-81937