Learn More
Medchemexpress LLC N-Boc-PEG10-alcohol | 2238844-74-9 | 98.0% | 557.67 g/mol | C25H51NO12 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW19085750MG
Encompass_Preferred
N-Boc-PEG10-alcohol is a polyethylene glycol (PEG)-based linker bearing a tert-butoxycarbonyl (Boc)-protected amine and a terminal hydroxyl. It is provided as a research reagent for use in medicinal chemistry workflows such as PROTAC assembly, bioconjugation, and linker optimization. Certificate of analysis and analytical data (HNMR/COA) are available on request; follow the product SDS for storage and handling.
Catalog No. 50-002-82244