Learn More
Medchemexpress LLC N-(Boc-PEG3)-N-bis(PEG2-alcohol) | 2055042-60-7 | C25H52N2O11 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14125450MG
Encompass_Preferred
N-(Boc-PEG3)-N-bis(PEG2-alcohol) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds that contain two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase and the other is for the target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-56044