Learn More
Medchemexpress LLC N-Boc-PEG5-alcohol | 1404111-67-6 | 97.0% | C15H31NO7 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1411911G
Encompass_Preferred
N-Boc-PEG5-alcohol is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins, by connecting an E3 ubiquitin ligase ligand and a target protein ligand via a linker.
  • PEG-based linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-55624