Learn More
Medchemexpress LLC N-Boc-PEG5-alcohol | 1404111-67-6 | 97.0% | C15H31NO7 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY141191
Encompass_Preferred
N-Boc-PEG5-alcohol is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs.
  • PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein.
  • PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
Catalog No. 50-003-52780