Learn More
Medchemexpress LLC N-Boc-PEG5-alcohol | 1404111-67-6 | 97.0% | C15H31NO7 | 5 G
SDP
Supplier:  Medchemexpress LLC HY1411915G
Encompass_Preferred
N-Boc-PEG5-alcohol is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs utilize two distinct ligands connected by a linker: one targets an E3 ubiquitin ligase, and the other targets a specific protein, enabling the selective degradation of target proteins via the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Suitable for synthesizing PROTACs
  • Utilizes the ubiquitin-proteasome system for protein degradation
  • For research use only
Catalog No. 50-003-53826