Learn More
Medchemexpress LLC N-Boc-PEG5-bromide | 1392499-32-9 | MFCD31536756 | 98.0% | 400.31 g/mol | C15H30BrNO6 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY120702500MG
Encompass_Preferred
N-Boc-PEG5-bromide is a Boc-protected polyethylene glycol (PEG5) alkyl bromide linker used as a building block in medicinal chemistry for PROTAC and ADC linker synthesis. It provides an orthogonally protected amine and a terminal bromide for downstream coupling and substitution reactions. Supplied with high reported purity, it is intended for research use under appropriate laboratory conditions.
  • Boc-protected amine for orthogonal deprotection.
  • Five-unit PEG spacer provides flexibility and solubility.
  • Terminal bromide enables nucleophilic substitution (SN2) chemistry.
  • Reported purity approximately 98.0%.
  • Suitable for assembly of PROTAC and ADC linkers.
  • Recommended cold storage to preserve stability.
Catalog No. 50-002-03454