Learn More
Medchemexpress LLC N-Boc-PEG7-alcohol | 1292268-13-3 | MFCD29049415 | 97.0% | 425.51 g/mol | C19H39NO9 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1305051G
Encompass_Preferred
N-Boc-PEG7-alcohol is a Boc-protected polyethylene glycol (PEG) linker with a terminal hydroxyl group, used as a flexible spacer in small-molecule conjugation chemistries such as PROTAC and ADC synthesis. It improves solubility and provides a site for further functionalization.
  • Boc-protected amino terminus for selective deprotection.
  • Terminal hydroxyl group suitable for further functionalization.
  • Peg7 spacer provides flexibility and improved solubility.
  • High purity suitable for research applications (97.0%).
  • Molecular weight approximately 425.5 g/mol.
Catalog No. 50-002-02144