Learn More
Medchemexpress LLC N-Boc-PEG9-alcohol | 2112731-44-7 | MFCD30723250 | 97.0% | 513.62 g·mol-1 | C23H47NO11 | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW071583250MG
Encompass_Preferred
N-Boc-PEG9-alcohol is a Boc-protected polyethylene glycol (PEG9) derivative with a terminal alcohol useful as a flexible, hydrophilic linker in PROTAC and antibody-drug conjugate (ADC) synthesis. It provides a cleavable spacer between conjugated molecular components and aids solubility and linker flexibility.
  • Provides a Boc-protected amine with a terminal alcohol functional group.
  • Contains a PEG9 chain for increased hydrophilicity and flexibility.
  • Suitable for use as a cleavable linker in PROTAC and ADC synthesis.
  • Used to introduce a flexible spacer between ligands or payloads.
  • Available in small research quantities for synthetic applications.
Catalog No. 50-002-03036