Learn More
Medchemexpress LLC N-Boc-PEG9-alcohol | 2112731-44-7 | MFCD30723250 | 97.0% | 513.62 g/mol | C23H47NO11 | 500 MG

Supplier: Medchemexpress LLC HYW071583500MG
N-Boc-PEG9-alcohol is a Boc-protected polyethylene glycol (PEG9) alcohol linker used as a spacer in conjugation chemistry, including ADC and PROTAC synthesis. It provides a PEG9 chain with a terminal hydroxyl group and a Boc-protected amine suitable for multi-step synthetic transformations.
- Provides a Boc-protected amine and terminal alcohol.
- Contains a nine-unit PEG spacer for solubility and flexibility.
- Molecular weight 513.62 g/mol and formula C23H47NO11.
- Typical analytical purity reported ≈97.0% depending on supplier.
- Suitable for linker incorporation in ADC, PROTAC, and other bioconjugation workflows.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.