Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Boc-PEG9-alcohol | 2112731-44-7 | MFCD30723250 | 98.0% | 513.62 g·mol⁻1 | C23H47NO11 | 5 G
SDP

Supplier:  Medchemexpress LLC HYW0715835G

Encompass_Preferred

N-Boc-PEG9-alcohol is a Boc-protected polyethylene glycol (PEG) linker with a primary alcohol at the PEG9 length, used as a cleavable spacer or linker in medicinal chemistry applications such as PROTAC and ADC synthesis.

  • Boc-protected primary amine functionality and a terminal alcohol.
  • Molecular weight approximately 513.62 g·mol⁻1.
  • Chemical formula C23H47NO11.
  • Reported purity ~98.0%.

Catalog No. 50-002-03120


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.