Learn More
Medchemexpress LLC N-Boc-piperazine | 57260-71-6 | 99.99% | 186.25 | 500 G
Supplier: Medchemexpress LLC HY30105500G
N-Boc-piperazine is an Alkyl/ether-based PROTAC linker. It can be utilized in the synthesis of PROTAC PD-1/PD-L1 degrader-1. The product is intended for research use only and is not sold to patients. It appears as a solid with a color ranging from white to light yellow. For shipping, it is kept at room temperature within the continental US, with potential variations elsewhere. For storage, the powder form should be kept at -20°C for 3 years or 4°C for 2 years. If in solvent, it should be stored at -80°C for 6 months or -20°C for 1 month.
- Alkyl/ether-based PROTAC linker.
- Can be used in the synthesis of PROTAC PD-1/PD-L1 degrader-1.
- High purity (99.99%).
- Available in various sizes.