Learn More
Medchemexpress LLC N-Boc-Tris | 146651-71-0 | ≥97.0% | C9H19NO5 | 50 G
SDP
Supplier:  Medchemexpress LLC HY14125350G
Encompass_Preferred
N-Boc-Tris is a protective reagent that utilizes the Boc protecting group to protect amino groups in organic synthesis and pharmaceutical research and development, preventing unnecessary side reactions. It is for research use only.
  • Protective reagent
  • Utilizes Boc protecting group
  • Protects amino groups in organic synthesis
  • Protects amino groups in pharmaceutical research and development
  • Avoids unnecessary side reactions
  • For research use only
Catalog No. 50-003-53193