Learn More
Medchemexpress LLC N-Butanoyl-DL-homoserine lactone | 98426-48-3 | 98.0% | 171.19 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11376410MG
Encompass_Preferred
N-Butanoyl-DL-homoserine lactone ((Rac)-C4-HSL) is a racemic mixture of N-Butanoyl-D-homoserine lactone and N-Butanoyl-L-homoserine lactone. N-Butanoyl-L-homoserine lactone is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). It possesses antibacterial activity and is utilized in antibacterial biofilm applications. N-Butanoyl-L-homoserine lactone aptamers are capable of blocking quorum sensing and inhibiting biofilm formation in Pseudomonas aeruginosa.
  • Racemic mixture of N-Butanoyl-D-homoserine lactone and N-Butanoyl-L-homoserine lactone.
  • Cleavable ADC linker used in synthesis of antibody-drug conjugates (ADCs).
  • Possesses antibacterial activity.
  • Utilized in antibacterial biofilm applications.
  • Aptamers capable of blocking quorum sensing.
  • Inhibits biofilm formation in Pseudomonas aeruginosa.
Catalog No. 50-004-37166