Learn More
Medchemexpress LLC N-Butanoyl-DL-homoserine lactone | 98426-48-3 | 98.0% | 171.19 | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HY11376410MM/1ML
Encompass_Preferred
N-Butanoyl-DL-homoserine lactone ((Rac)-C4-HSL) is a racemic mixture of N-Butanoyl-D-homoserine lactone and N-Butanoyl-L-homoserine lactone. This compound is used as a cleavable ADC linker in the synthesis of antibody-drug conjugates.
  • Exhibits antibacterial activity
  • Used in antibacterial biofilm applications
  • Aptamers block quorum sensing
  • Inhibits biofilm formation in Pseudomonas aeruginosa
Catalog No. 50-004-37359