Learn More
Medchemexpress LLC N-butanoyl-DL-homoserine lactone | 98426-48-3 | MFCD01862909 | 98.0% | 171.19 g/mol | C8H13NO3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1137645MG
Encompass_Preferred
N-butanoyl-DL-homoserine lactone is a racemic N-acyl-homoserine lactone used as an analytical standard and research reagent to study bacterial quorum sensing, biofilm formation, and related signaling pathways. Supplied as a solid or as a DMSO solution at high purity, it is appropriate for analytical method development and biological assays.
  • Racemic homoserine lactone standard for quorum sensing research.
  • High purity (98.0%) suitable for analytical method development (HPLC/GC/MS).
  • Available as a solid or as a 10 mM solution in DMSO.
  • Stable when stored as powder at -20°C or 4°C according to datasheet.
  • Suitable for biological assays and synthetic chemistry applications.
Catalog No. 50-002-64395