Learn More
AdipoGen N-Butanoyl-DL-homoserine lactone (5 mg)
SDP
Supplier:  AdipoGen CDXB0280M005
Encompass_Preferred
N-Butanoyl-DL-homoserine lactone (5 mg), AdipoGen Life Sciences. Chemical. CAS: 98426-48-3. Formula: C8H13NO3. MW: 171.2. Synthetic. N-Butyryl-DL-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones (AHL) are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms.
Catalog No. 50-149-0269