Learn More
Medchemexpress LLC N-butyl-4-[(phenylamino)sulfonyl]-N-(phenylmethyl)-benzamide | 2244678-29-1 | 98.4% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY12263010MM 1ML
Encompass_Preferred
TH-257 is a potent, selective allosteric inhibitor of LIM kinase 1 and LIM kinase 2 used as a chemical probe in cell biology research. It is supplied as a 10 mM solution in DMSO (1 mL vial), with typical purity of 98.36% and a molecular weight of 422.54 g/mol.
  • Potent inhibition of LIMK1 and LIMK2 (IC50 84 nM and 39 nM).
  • Allosteric mode of action with high selectivity against the kinome.
  • Provided as a ready-to-use 10 mM solution in DMSO for in vitro studies.
  • High purity suitable for research applications.
  • Molecular formula C24H26N2O3S and molecular weight 422.54 g/mol.
Catalog No. 50-002-34775