Learn More
Medchemexpress LLC N-butylthiophosphoric triamide | 94317-64-3 | MFCD00269941 | 99.8% | 167.22 g/mol | C4H14N3PS | 25 G
SDP
Supplier:  Medchemexpress LLC HY10719925G
Encompass_Preferred
N-Butylthiophosphoric triamide (NBPT) is an organophosphorus urease inhibitor that reduces the enzymatic conversion of urea to ammonia, improving nitrogen fertilizer efficiency and lowering ammonia volatilization. It is used in agricultural research and fertilizer stabilization studies and is supplied in solid and solution formats for formulation and laboratory use.
  • Inhibits urease activity to reduce ammonia loss.
  • Improves nitrogen use efficiency in fertilizer applications.
  • Available as solid and concentrated solution formats for flexibility.
  • Supplied with safety and analytical documentation such as SDS and COA.
  • Suitable for agricultural research and fertilizer formulation studies.
Catalog No. 50-002-40839