Learn More
Strem, An Ascensus Company CAS# 1118-46-3. 250g. n-Butyltin trichloride, min. 95%. MFCD00000515
SDP
Supplier:  Strem, An Ascensus Company 935004250G
Encompass
CAS# 1118-46-3. 250g. n-Butyltin trichloride, min. 95%. MFCD00000515. Molecular Weight: 282.17. Molecular Formula: C4H9SnCl3. Color/form: colorless to yellow liq. Strem# 93-5004. http://www.strem.com/catalog/v/93-5004/
Catalog No. 50-901-15543