Learn More
Apexbio Technology LLC N-Carbamyl-L-glutamic acid 1188-38-1 100mg
SDP
Supplier:  Apexbio Technology LLC B7907100
Encompass_Preferred
N-Carbamyl-L-glutamic acid is a small-molecule activator targeting mitochondrial carbamoyl phosphate synthetase 1 (CPS1) It is designed to stimulate CPS1 activity thereby enhancing carbamoyl phosphate formation and promoting ammonia detoxification via the urea cycle N-Carbamyl-L-glutamic acid exerts its biological activity primarily through activation of CPS1 In experimental models N-carbamyl-L-glutamic acid enhances ammonia clearance and suppresses cellular proliferation across multiple cancer cell lines exhibiting antiproliferative effects at nanomolar concentrations Based on these pharmacological properties N-carbamyl-L-glutamic acid holds research potential in studies of hyperammonemia-related metabolic disturbances and cancer biology
Catalog No. 50-000-88136