Learn More
Medchemexpress LLC N-carbobenzoxy-D-homoserine | 41088-85-1 | 253.25 g·mol⁻¹ | C12H15NO5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW025723A100MG
Encompass_Preferred
N-Carbobenzoxy-D-homoserine is D-homoserine bearing an N-carbobenzoxy (benzyloxycarbonyl, Cbz) protecting group. It is used as a synthetic intermediate in medicinal chemistry and for preparation of antibody-drug conjugates (ADCs). Solid material; molecular formula C12H15NO5, molecular weight 253.25 g·mol⁻¹.
  • Cbz-protected D-homoserine for peptide and small-molecule synthesis.
  • Useful as a drug intermediate in ADC linker or payload assembly.
  • Supplied as a solid with defined molecular formula and molecular weight.
  • Recommended storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years).
  • Available in small research pack sizes suitable for laboratory use.
Catalog No. 50-003-01458