Learn More
Medchemexpress LLC N-carbobenzoxy-DL-norvaline | 21691-43-0 | MFCD00020434 | 98.0% | 251.28 g/mol | C13H17NO4 | 10 G

Supplier: Medchemexpress LLC HYW14212610G
N-carbobenzoxy-DL-norvaline is an amino acid derivative of valine supplied as a white to off-white solid for research use. It is used in biochemical and pharmacological studies related to amino acid metabolism and ergogenic effects, and the product page provides storage and handling guidance.
- Derivative of the amino acid valine.
- Supplied as a white to off-white solid.
- Molecular formula C13H17NO4; molecular weight 251.28 g/mol.
- CAS number 21691-43-0 for unambiguous identification.
- Storage: powder -20°C for 3 years, 4°C for 2 years; in solvent -80°C for 6 months, -20°C for 1 month.
- Available in small pack sizes such as 5 G, 10 G, and 25 G; bulk options available.
- Intended for research use only; not for human consumption.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.