Learn More
Medchemexpress LLC Nε-(carboxymethyl)-L-lysine | 5746-04-3 | MFCD04114278 | 95.0% | 204.22 g/mol | C8H16N2O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW2867435MG
Encompass_Preferred
Nε-(Carboxymethyl)-L-lysine is a carboxymethylated derivative of lysine used as an analytical standard and biomarker reagent. It is supplied as a white to off-white solid and is commonly used in studies of advanced glycation end products and in analytical workflows such as mass spectrometry and chromatography.
  • White to off-white solid suitable for analytical use.
  • Used as an analytical standard for advanced glycation end product research.
  • Compatible with LC-MS and chromatographic methods.
  • Typical purity around 95.0%, with higher-purity batches available.
  • Powder stable at -20 °C for long-term storage; solutions require colder storage.
Catalog No. 50-002-80545