Learn More
Medchemexpress LLC N-Cbz-glycyl-glycyl-D-phenylalanine | 1802522-76-4 | 99.2% | 413.42 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY48321
Encompass_Preferred
N-Cbz-glycyl-glycyl-D-phenylalanine is a cleavable ADC linker.
  • High purity
  • Off-white to light yellow solid
  • Store at 4°C, sealed, away from moisture
  • Used as an antibody-drug conjugate linker
Catalog No. 50-003-66112