Learn More
Medchemexpress LLC N-Cbz-hydroxy-L-proline | 13504-85-3 | MFCD00037329 | 99.9% | 265.26 | C13H15NO5 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01604210G
Encompass_Preferred
N-Cbz-hydroxy-L-proline is a Cbz-protected derivative of hydroxy-L-proline supplied as a white to off-white solid for research use only. It is used as a building block in organic synthesis and as a reagent in biochemical studies; consult supplier documents for solubility, storage, and handling recommendations.
  • High chemical purity (99.9%).
  • C13H15NO5 formula; molecular weight 265.26 g/mol.
  • Cbz-protected hydroxyproline useful as a synthetic building block.
  • Solid form, white to off-white, easy to weigh and handle.
  • Stable under recommended storage: powder -20 °C (3 years) or 4 °C (2 years).
  • Follow DMSO handling guidance to avoid hygroscopic solubility issues.
Catalog No. 50-002-99938