Learn More
Medchemexpress LLC N-Cbz-L-valine | 1149-26-4 | 99.8% | 251.28 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW008753
Encompass_Preferred
Z-L-Val-OH (N-Cbz-L-valine) is an amino acid derivative and a key intermediate in organic synthesis. It can be utilized in the synthesis of anti-cytomegalovirus compounds, as well as peptide inhibitors of human spleen plasminogen activator (SFP) and human leukocyte elastase-like protease (ELP).
  • Amino acid derivative
  • Key intermediate in organic synthesis
  • Utilized in the synthesis of anti-cytomegalovirus compounds
  • Used in the synthesis of peptide inhibitors of human spleen plasminogen activator (SFP)
  • Used in the synthesis of human leukocyte elastase-like protease (ELP) inhibitors
Catalog No. 50-004-51314