Learn More
TARGETMOL CHEMICALS INC N-CBZ-Phe-Arg-AMC hydrochlorid
SDP
Supplier:  TARGETMOL CHEMICALS INC TP1013L25MG
Encompass_Preferred
Also available in 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. N-CBZ-Phe-Arg-AMC hydrochloride (N-CBZ-Phe-Arg-AMC hydrochloride (65147-22-0 Free base)) is a cysteine peptidase and serine protease substrate that can be used to assess cysteine peptidase and serine protease activity. Purity 99.99%
Catalog No. 50-288-0418