Learn More
AB Sciex LLC N-CHO Capillary for N-Linked Oligosaccharide Profiling
Supplier: AB Sciex LLC 477601
N-CHO (PVA) Capillary with laser burned window - 50 µm ID x 65 cm total length. Capillary for use in PA 800 series and P/ACE MDQ Capillary Cartridges. This capillary is also used with Carbohydrate Labeling and Analysis Kit (part number 477600). Capillaries must be trimmed to optimize speed/resolution.