Learn More
Cayman Chemical N-Cholyl-L-LeucIn 100mg
SDP
Supplier:  Cayman Chemical 40930100 MG
Encompass_Preferred
A bile acid amidate; has been found in fecal samples from extremely premature infants with mild or severe brain injuries; has been used as a standard for the detection of endogenous bile acid amidates in human plasma and fecal samples
Catalog No. 50-294-3596